gRNA Design Details
- Target DNA Sequence
- ATCCCCGCAATTAACATAAG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 147899806 - 147899828 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.32/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ACVR2A
- Entrez Gene ID
-
92
- Gene Name
- activin A receptor type 2A
- Gene Aliases
-
ACTRII, ACVR2
- Chromosome Location
-
Chr.2: 147844517 - 147930827 on GRCh38
- UniGene ID
- Hs.470174
- Species
-
Human