gRNA Design Details
- Target DNA Sequence
- CCATTAGCGCATCACAGTCG
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 36684194 - 36684216 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
97.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CDKN1A
- Entrez Gene ID
-
1026
- Gene Name
- cyclin dependent kinase inhibitor 1A
- Gene Aliases
-
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
- Chromosome Location
-
Chr.6: 36676460 - 36687339 on GRCh38
- UniGene ID
- Hs.370771
- Species
-
Human