gRNA Design Details
- Target DNA Sequence
- CTTTGTCACCGAGACACCAC
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 36684257 - 36684279 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CDKN1A
- Entrez Gene ID
-
1026
- Gene Name
- cyclin dependent kinase inhibitor 1A
- Gene Aliases
-
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
- Chromosome Location
-
Chr.6: 36676460 - 36687339 on GRCh38
- UniGene ID
- Hs.370771
- Species
-
Human