gRNA Design Details
- Target DNA Sequence
- GCTACTACCATTATTGCTGG
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 45076744 - 45076766 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Kazald1
- Entrez Gene ID
-
107250
- Gene Name
- Kazal-type serine peptidase inhibitor domain 1
- Gene Aliases
-
AI842353, Bono1, IGFBP-rP10
- Chromosome Location
-
Chr.19: 45074485 - 45079289 on GRCm38
- UniGene ID
- Mm.24417
- Species
-
Mouse