gRNA Design Details
- Target DNA Sequence
- TCCAAGCCTTGTCTTAATGG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 34614549 - 34614571 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Egfl8
- Entrez Gene ID
-
81701
- Gene Name
- EGF-like domain 8
- Gene Aliases
-
Ng3
- Chromosome Location
-
Chr.17: 34613200 - 34616572 on GRCm38
- UniGene ID
- Mm.231343
- Species
-
Mouse