gRNA Design Details
- Target DNA Sequence
- CGTAGGCCTCGTAAACCTCA
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 227817239 - 227817261 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.29/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS38
- Entrez Gene ID
-
339501
- Gene Name
- protease, serine 38
- Gene Aliases
-
MPN2
- Chromosome Location
-
Chr.1: 227815708 - 227846470 on GRCh38
- UniGene ID
- Hs.97604
- Species
-
Human