gRNA Design Details

Target DNA Sequence
ATTGTACCACCACCTAGGCA
PAM Sequence
TGG
Target locus
Chr.19: 38283669 - 38283691 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.01/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPINT2
Entrez Gene ID
10653
Gene Name
serine peptidase inhibitor, Kunitz type 2
Gene Aliases
DIAR3, HAI-2, HAI2, Kop, PB
Chromosome Location
Chr.19: 38264458 - 38292614 on GRCh38
UniGene ID
Hs.31439
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_021102.3 NP_066925.1 2