gRNA Design Details

Target DNA Sequence
GGAATCCGCTGCATTCCTGC
PAM Sequence
TGG
Target locus
Chr.19: 38287900 - 38287922 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 85.88/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPINT2
Entrez Gene ID
10653
Gene Name
serine peptidase inhibitor, Kunitz type 2
Gene Aliases
DIAR3, HAI-2, HAI2, Kop, PB
Chromosome Location
Chr.19: 38264458 - 38292614 on GRCh38
UniGene ID
Hs.31439
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001166103.1 NP_001159575.1 2
NM_021102.3 NP_066925.1 3