gRNA Design Details
- Target DNA Sequence
- TGACAGTGGCACATTTCTTG
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 38283772 - 38283794 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINT2
- Entrez Gene ID
-
10653
- Gene Name
- serine peptidase inhibitor, Kunitz type 2
- Gene Aliases
-
DIAR3, HAI-2, HAI2, Kop, PB
- Chromosome Location
-
Chr.19: 38264458 - 38292614 on GRCh38
- UniGene ID
- Hs.31439
- Species
-
Human