gRNA Design Details

Target DNA Sequence
GATGCATGTCGGCTTCTCGC
PAM Sequence
TGG
Target locus
Chr.2: 219246756 - 219246778 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.06/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK16
Entrez Gene ID
8576
Gene Name
serine/threonine kinase 16
Gene Aliases
KRCT, MPSK, PKL12, TSF1
Chromosome Location
Chr.2: 219245469 - 219250337 on GRCh38
UniGene ID
Hs.153003
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001008910.2 NP_001008910.1 3
XM_011512067.2 XP_011510369.1 3
XM_017005160.1 XP_016860649.1 3