gRNA Design Details

Target DNA Sequence
CAAAACCACTCGAATAAGTC
PAM Sequence
AGG
Target locus
Chr.22: 49920169 - 49920191 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.42/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CRELD2
Entrez Gene ID
79174
Gene Name
cysteine rich with EGF like domains 2
Gene Aliases
-
Chromosome Location
Chr.22: 49918630 - 49927540 on GRCh38
UniGene ID
Hs.211282
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001135101.2 NP_001128573.1 4
NM_001284317.1 NP_001271246.1 4
NM_001284318.1 NP_001271247.1 4
NM_024324.4 NP_077300.3 4
NR_104295.1 4
XM_005261737.2 XP_005261794.1 4
XM_005261738.4 XP_005261795.1 4
XM_011530372.2 XP_011528674.1 4
XM_017028938.1 XP_016884427.1 4