Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_002039.3 | NP_002030.2 | 2 |
NM_207123.2 | NP_997006.1 | 2 | |
XM_006714167.3 | XP_006714230.1 | 2 | |
XM_006714168.3 | XP_006714231.1 | 2 | |
XM_017007966.1 | XP_016863455.1 | 3 | |
XM_017007967.1 | XP_016863456.1 | 2 | |
XM_017007968.1 | XP_016863457.1 | 3 | |
XM_017007969.1 | XP_016863458.1 | 2 | |
XM_017007970.1 | XP_016863459.1 | 2 | |
XM_017007971.1 | XP_016863460.1 | 2 |