gRNA Design Details
- Target DNA Sequence
- TAGTGGTTGACGAACTCGGT
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 51913278 - 51913300 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ACVRL1
- Entrez Gene ID
-
94
- Gene Name
- activin A receptor like type 1
- Gene Aliases
-
ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I
- Chromosome Location
-
Chr.12: 51906913 - 51923361 on GRCh38
- UniGene ID
- Hs.591026
- Species
-
Human