gRNA Design Details
- Target DNA Sequence
- GATGCAGTACGAGGACTGGA
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 38750017 - 38750039 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRSF7
- Entrez Gene ID
-
6432
- Gene Name
- serine and arginine rich splicing factor 7
- Gene Aliases
-
9G8, AAG3, SFRS7
- Chromosome Location
-
Chr.2: 38743599 - 38751494 on GRCh38
- UniGene ID
- Hs.309090
- Species
-
Human