gRNA Design Details
- Target DNA Sequence
- AAATTTAGCACTTCCTCCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 122453786 - 122453808 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IL2
- Entrez Gene ID
-
3558
- Gene Name
- interleukin 2
- Gene Aliases
-
IL-2, TCGF, lymphokine
- Chromosome Location
-
Chr.4: 122449479 - 122456495 on GRCh38
- UniGene ID
- Hs.89679
- Species
-
Human