gRNA Design Details
- Target DNA Sequence
- CAACCTGCCAGACGGAAAAC
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 78492802 - 78492824 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PTGFR
- Entrez Gene ID
-
5737
- Gene Name
- prostaglandin F receptor
- Gene Aliases
-
FP
- Chromosome Location
-
Chr.1: 78490974 - 78540701 on GRCh38
- UniGene ID
- Hs.654365
- Species
-
Human