gRNA Design Details

Target DNA Sequence
GATCATCTCCTTCCTGAAGG
PAM Sequence
AGG
Target locus
Chr.2: 241030298 - 241030320 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 87.43/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SNED1
Entrez Gene ID
25992
Gene Name
sushi, nidogen and EGF like domains 1
Gene Aliases
SST3, Snep
Chromosome Location
Chr.2: 240997409 - 241095568 on GRCh38
UniGene ID
Hs.471834
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001080437.1 NP_001073906.1 2
XM_011510931.2 XP_011509233.1 3
XM_011510932.2 XP_011509234.1 3
XM_011510933.2 XP_011509235.1 3
XM_011510934.2 XP_011509236.1 3
XM_011510937.2 XP_011509239.1 3
XM_017003773.1 XP_016859262.1 3
XM_017003774.1 XP_016859263.1 2
XM_017003775.1 XP_016859264.1 3
XM_017003776.1 XP_016859265.1 3
XM_017003777.1 XP_016859266.1 3
XM_017003778.1 XP_016859267.1 2
XM_017003779.1 XP_016859268.1 3
XM_017003780.1 XP_016859269.1 2
XM_017003781.1 XP_016859270.1 2