Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 2 |
XM_011510931.2 | XP_011509233.1 | 3 | |
XM_011510932.2 | XP_011509234.1 | 3 | |
XM_011510933.2 | XP_011509235.1 | 3 | |
XM_011510934.2 | XP_011509236.1 | 3 | |
XM_011510937.2 | XP_011509239.1 | 3 | |
XM_017003773.1 | XP_016859262.1 | 3 | |
XM_017003774.1 | XP_016859263.1 | 2 | |
XM_017003775.1 | XP_016859264.1 | 3 | |
XM_017003776.1 | XP_016859265.1 | 3 | |
XM_017003777.1 | XP_016859266.1 | 3 | |
XM_017003778.1 | XP_016859267.1 | 2 | |
XM_017003779.1 | XP_016859268.1 | 3 | |
XM_017003780.1 | XP_016859269.1 | 2 | |
XM_017003781.1 | XP_016859270.1 | 2 |