gRNA Design Details

Target DNA Sequence
TGTCTGTGGCTCAACAGACA
PAM Sequence
AGG
Target locus
Chr.20: 916137 - 916159 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.61/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
Chromosome Location
Chr.20: 869899 - 916367 on GRCh38
UniGene ID
Hs.278973
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001322809.1 NP_001309738.1 1
NM_015985.3 NP_057069.1 1
XM_011529239.2 XP_011527541.1 1