gRNA Design Details
- Target DNA Sequence
- GCAAGAAGGTGTAGCTACAG
- PAM Sequence
- TGG
- Target locus
-
Chr.20: 916070 - 916092 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ANGPT4
- Entrez Gene ID
-
51378
- Gene Name
- angiopoietin 4
- Gene Aliases
-
ANG3, ANG4
- Chromosome Location
-
Chr.20: 869899 - 916367 on GRCh38
- UniGene ID
- Hs.278973
- Species
-
Human