gRNA Design Details

Target DNA Sequence
CTTCTCGTCCAAGAACTACG
PAM Sequence
TGG
Target locus
Chr.9: 110579144 - 110579166 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 95.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SVEP1
Entrez Gene ID
79987
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
C9orf13, CCP22, POLYDOM, SEL-OB, SELOB
Chromosome Location
Chr.9: 110365248 - 110579880 on GRCh38
UniGene ID
Hs.522334
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_153366.3 NP_699197.3 1