gRNA Design Details
- Target DNA Sequence
- CTTGGACGAGAAGGTCACGA
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 110579154 - 110579176 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SVEP1
- Entrez Gene ID
-
79987
- Gene Name
- sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
- Gene Aliases
-
C9orf13, CCP22, POLYDOM, SEL-OB, SELOB
- Chromosome Location
-
Chr.9: 110365248 - 110579880 on GRCh38
- UniGene ID
- Hs.522334
- Species
-
Human