gRNA Design Details
- Target DNA Sequence
- TCCAAGCAAGTTGTAGCTCA
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 21077786 - 21077808 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IFNB1
- Entrez Gene ID
-
3456
- Gene Name
- interferon beta 1
- Gene Aliases
-
IFB, IFF, IFN-beta, IFNB
- Chromosome Location
-
Chr.9: 21077105 - 21077963 on GRCh38
- UniGene ID
- Hs.93177
- Species
-
Human