gRNA Design Details
- Target DNA Sequence
- TCTCAACAACAAGTACTCGG
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 127520354 - 127520376 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Shmt2
- Entrez Gene ID
-
108037
- Gene Name
- serine hydroxymethyltransferase 2 (mitochondrial)
- Gene Aliases
-
2700043D08Rik, AA408223, AA986903
- Chromosome Location
-
Chr.10: 127517123 - 127522444 on GRCm38
- UniGene ID
- Mm.29890
- Species
-
Mouse