gRNA Design Details

Target DNA Sequence
ATCCAGGTCAAAGGCTTCCA
PAM Sequence
AGG
Target locus
Chr.12: 57231769 - 57231791 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 65.37/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SHMT2
Entrez Gene ID
6472
Gene Name
serine hydroxymethyltransferase 2
Gene Aliases
GLYA, HEL-S-51e, SHMT
Chromosome Location
Chr.12: 57229573 - 57234935 on GRCh38
UniGene ID
Hs.741179
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001166356.1 NP_001159828.1 4
NM_001166357.1 NP_001159829.1 4
NM_001166358.1 NP_001159830.1 4
NM_001166359.1 NP_001159831.1 4
NM_005412.5 NP_005403.2 4
NR_029415.1 3
NR_029416.1 3
NR_029417.1 3
NR_048562.1 3
XM_011538675.2 XP_011536977.1 4
XM_011538676.2 XP_011536978.1 4
XM_011538677.2 XP_011536979.1 3
XM_011538678.2 XP_011536980.1 3