gRNA Design Details
- Target DNA Sequence
- GCTGAATATCAGCGCGGACA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 101239086 - 101239108 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- S1PR1
- Entrez Gene ID
-
1901
- Gene Name
- sphingosine-1-phosphate receptor 1
- Gene Aliases
-
CD363, CHEDG1, D1S3362, ECGF1, EDG-1, EDG1, S1P1
- Chromosome Location
-
Chr.1: 101236749 - 101241520 on GRCh38
- UniGene ID
- Hs.154210
- Species
-
Human