gRNA Design Details
- Target DNA Sequence
- CGTTGGCAGTCCGCCTTAAC
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 116941941 - 116941963 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
93.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PTGFRN
- Entrez Gene ID
-
5738
- Gene Name
- prostaglandin F2 receptor inhibitor
- Gene Aliases
-
CD315, CD9P-1, EWI-F, FPRP, SMAP-6
- Chromosome Location
-
Chr.1: 116909923 - 116990358 on GRCh38
- UniGene ID
- Hs.418093
- Species
-
Human