gRNA Design Details

Target DNA Sequence
ATATCCGCAACCAGCACCGC
PAM Sequence
TGG
Target locus
Chr.14: 105051813 - 105051835 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 92.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GPR132
Entrez Gene ID
29933
Gene Name
G protein-coupled receptor 132
Gene Aliases
G2A
Chromosome Location
Chr.14: 105049389 - 105065550 on GRCh38
UniGene ID
Hs.532504
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001278694.1 NP_001265623.1 5
NM_001278695.1 NP_001265624.1 3
NM_001278696.1 NP_001265625.1 2
NM_013345.3 NP_037477.1 4