gRNA Design Details
- Target DNA Sequence
- CGGATATAGATGACCCAGAG
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 105051829 - 105051851 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GPR132
- Entrez Gene ID
-
29933
- Gene Name
- G protein-coupled receptor 132
- Gene Aliases
-
G2A
- Chromosome Location
-
Chr.14: 105049389 - 105065550 on GRCh38
- UniGene ID
- Hs.532504
- Species
-
Human