gRNA Design Details
- Target DNA Sequence
- GAGTCCTGGCCATCCAGTTC
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 35759879 - 35759901 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.71/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PROSER3
- Entrez Gene ID
-
148137
- Gene Name
- proline and serine rich 3
- Gene Aliases
-
C19orf55
- Chromosome Location
-
Chr.19: 35758139 - 35770115 on GRCh38
- UniGene ID
- Hs.527982
- Species
-
Human