gRNA Design Details
- Target DNA Sequence
- ACTAACCATTATTGCTATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 161346188 - 161346210 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPTSSB
- Entrez Gene ID
-
165679
- Gene Name
- serine palmitoyltransferase small subunit B
- Gene Aliases
-
ADMP, C3orf57, SSSPTB
- Chromosome Location
-
Chr.3: 161344784 - 161373123 on GRCh38
- UniGene ID
- Hs.369104
- Species
-
Human