gRNA Design Details
- Target DNA Sequence
- GAGGACCTGTTCTACAAGTA
- PAM Sequence
- CGG
- Target locus
-
Chr.12: 120469497 - 120469519 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRSF9
- Entrez Gene ID
-
8683
- Gene Name
- serine and arginine rich splicing factor 9
- Gene Aliases
-
SFRS9, SRp30c
- Chromosome Location
-
Chr.12: 120461668 - 120469755 on GRCh38
- UniGene ID
- Hs.706889
- Species
-
Human