gRNA Design Details
- Target DNA Sequence
- ATGGATTCGGGATTGCTCTG
- PAM Sequence
- AGG
- Target locus
-
Chr.20: 45512603 - 45512625 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.75/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINT3
- Entrez Gene ID
-
10816
- Gene Name
- serine peptidase inhibitor, Kunitz type 3
- Gene Aliases
-
HKIB9
- Chromosome Location
-
Chr.20: 45512461 - 45515624 on GRCh38
- UniGene ID
- Hs.370724
- Species
-
Human