gRNA Design Details
- Target DNA Sequence
- AGGTCGGCGAGTTTGTGGTA
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 89758100 - 89758122 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RIPK2
- Entrez Gene ID
-
8767
- Gene Name
- receptor interacting serine/threonine kinase 2
- Gene Aliases
-
CARD3, CARDIAK, CCK, GIG30, RICK, RIP2
- Chromosome Location
-
Chr.8: 89757747 - 89791064 on GRCh38
- UniGene ID
- Hs.103755
- Species
-
Human