gRNA Design Details

Target DNA Sequence
TGCGAACGACAGGCACACAG
PAM Sequence
AGG
Target locus
Chr.10: 101062962 - 101062984 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 91.58/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
Chromosome Location
Chr.10: 101061141 - 101068141 on GRCh38
UniGene ID
Hs.534859
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001319303.1 NP_001306232.1 2
NM_030929.4 NP_112191.2 2
XM_005270194.3 XP_005270251.1 2
XM_017016714.1 XP_016872203.1 1
XM_017016715.1 XP_016872204.1 1