gRNA Design Details

Target DNA Sequence
GCATAAGAAAAAGCGGAGTT
PAM Sequence
CGG
Target locus
Chr.2: 202034934 - 202034956 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.22/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
Chromosome Location
Chr.2: 202034587 - 202038437 on GRCh38
UniGene ID
Hs.173859
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_003507.1 NP_003498.1 1