gRNA Design Details
- Target DNA Sequence
- GCATAAGAAAAAGCGGAGTT
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 202034934 - 202034956 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.22/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FZD7
- Entrez Gene ID
-
8324
- Gene Name
- frizzled class receptor 7
- Gene Aliases
-
FzE3
- Chromosome Location
-
Chr.2: 202034587 - 202038437 on GRCh38
- UniGene ID
- Hs.173859
- Species
-
Human