gRNA Design Details
- Target DNA Sequence
- ATGTTCTGTTCGCCACCACG
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 5494592 - 5494614 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NTF3
- Entrez Gene ID
-
4908
- Gene Name
- neurotrophin 3
- Gene Aliases
-
HDNF, NGF-2, NGF2, NT-3, NT3
- Chromosome Location
-
Chr.12: 5432114 - 5495299 on GRCh38
- UniGene ID
- Hs.99171
- Species
-
Human