gRNA Design Details
- Target DNA Sequence
- AGCCTACAAGTGCTTCTACC
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 55118643 - 55118665 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- KDR
- Entrez Gene ID
-
3791
- Gene Name
- kinase insert domain receptor
- Gene Aliases
-
CD309, FLK1, VEGFR, VEGFR2
- Chromosome Location
-
Chr.4: 55078259 - 55125595 on GRCh38
- UniGene ID
- Hs.479756
- Species
-
Human