gRNA Design Details
- Target DNA Sequence
- GTGTTTCTCTTGATCTGCCC
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 55121159 - 55121181 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.3/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- KDR
- Entrez Gene ID
-
3791
- Gene Name
- kinase insert domain receptor
- Gene Aliases
-
CD309, FLK1, VEGFR, VEGFR2
- Chromosome Location
-
Chr.4: 55078259 - 55125595 on GRCh38
- UniGene ID
- Hs.479756
- Species
-
Human