gRNA Design Details

Target DNA Sequence
GATGAGCTGATACCCAATGG
PAM Sequence
AGG
Target locus
Chr.16: 3850949 - 3850971 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.59/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
Chromosome Location
Chr.16: 3725054 - 3880727 on GRCh38
UniGene ID
Hs.459759
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001079846.1 NP_001073315.1 2
NM_004380.2 NP_004371.2 2
XM_005255124.4 XP_005255181.1 2
XM_005255125.4 XP_005255182.1 2
XM_006720848.3 XP_006720911.1 2
XM_011522382.2 XP_011520684.1 2
XM_017022944.1 XP_016878433.1 2