gRNA Design Details
- Target DNA Sequence
- TGTGATTTTGGGCCCAGGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 19158338 - 19158360 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
60.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GRAPL
- Entrez Gene ID
-
400581
- Gene Name
- GRB2 related adaptor protein like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 19123188 - 19159187 on GRCh38
- UniGene ID
- Hs.647426
- Species
-
Human