gRNA Design Details
- Target DNA Sequence
- CACGTATTTGAGCTGCACCT
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 219601300 - 219601322 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.8/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK11IP
- Entrez Gene ID
-
114790
- Gene Name
- serine/threonine kinase 11 interacting protein
- Gene Aliases
-
LIP1, LKB1IP, STK11IP1
- Chromosome Location
-
Chr.2: 219597851 - 219616451 on GRCh38
- UniGene ID
- Hs.22410
- Species
-
Human