gRNA Design Details
- Target DNA Sequence
- GTTAAGGTTGCCATTACAGC
- PAM Sequence
- CGG
- Target locus
-
Chr.20: 45723979 - 45724001 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.41/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINT4
- Entrez Gene ID
-
391253
- Gene Name
- serine peptidase inhibitor, Kunitz type 4
- Gene Aliases
-
C20orf137, SPINT3, dJ601O1.1
- Chromosome Location
-
Chr.20: 45722349 - 45725696 on GRCh38
- UniGene ID
- Hs.677891
- Species
-
Human