gRNA Design Details

Target DNA Sequence
CACCGACGTGTAGCAGCAGA
PAM Sequence
AGG
Target locus
Chr.9: 100473600 - 100473622 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.95/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TMEFF1
Entrez Gene ID
8577
Gene Name
transmembrane protein with EGF like and two follistatin like domains 1
Gene Aliases
C9orf2, CT120.1, H7365, TR-1
Chromosome Location
Chr.9: 100473238 - 100577636 on GRCh38
UniGene ID
Hs.598100
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_003692.4 NP_003683.2 1