gRNA Design Details
- Target DNA Sequence
- ATATTGGGACAGGGGAGCTT
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 109338390 - 109338412 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
83.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk33
- Entrez Gene ID
-
117229
- Gene Name
- serine/threonine kinase 33
- Gene Aliases
-
4921505G21Rik, AW061264
- Chromosome Location
-
Chr.7: 109279213 - 109439053 on GRCm38
- UniGene ID
- Mm.389950
- Species
-
Mouse