gRNA Design Details
- Target DNA Sequence
- TGGACAACAAGAGGTGTGAC
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 46743003 - 46743025 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS45
- Entrez Gene ID
-
377047
- Gene Name
- protease, serine 45
- Gene Aliases
-
TESSP5
- Chromosome Location
-
Chr.3: 46742091 - 46744755 on GRCh38
- UniGene ID
- Hs.667712
- Species
-
Human