Gene Transcripts

Gene Symbol
CEACAM8
Entrez Gene ID
1088
Gene Name
carcinoembryonic antigen related cell adhesion molecule 8
Gene Aliases
CD66b, CD67, CGM6, NCA-95
Chromosome Location
Chr. 19: 42580243 - 42595016 on Build GRCh38
UniGene ID
Hs.41
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001816.3 NP_001807.2 4 895
XM_011526340.2 XP_011524642.1 4 895
XM_017026194.1 XP_016881683.1 4 895
XM_017026195.1 XP_016881684.1 4 895
XM_017026196.1 XP_016881685.1 4 895
XM_017026197.1 XP_016881686.1 3 621
GenBank mRNA AK297012.1 - 4 584
AK312652.1 - 4 892
BC026263.1 - 4 863 4618311
D90064.1 - 4 879
M33326.1 - 4 879
X52378.1 - 4 825

View Full Product Details