Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003334.3 | NP_003325.2 | 23 | 3010 | |
NM_153280.2 | NP_695012.1 | 23 | 2938 | ||
XM_005272649.1 | XP_005272706.1 | 24 | 3388 | ||
XM_011543954.2 | XP_011542256.1 | 24 | 3030 | ||
XM_017029777.1 | XP_016885266.1 | 23 | 4105 | ||
XM_017029778.1 | XP_016885267.1 | 24 | 5279 | ||
XM_017029779.1 | XP_016885268.1 | 23 | 5787 | ||
XM_017029780.1 | XP_016885269.1 | 23 | 2895 | ||
XM_017029781.1 | XP_016885270.1 | 23 | 2943 | ||
GenBank mRNA | AF258566.1 | - | 2 | 1348 | |
AK097343.1 | - | 7 | 1910 | ||
AK131341.1 | - | 2 | 1134 | ||
AK293161.1 | - | 12 | 1602 | ||
AK296866.1 | - | 13 | 1628 | ||
BC013041.2 | - | 23 | 2877 | 3543021 | |
BC020261.1 | - | 20 | 2572 | ||
M58028.1 | - | 23 | 2916 | ||
X52897.1 | - | 16 | 2082 | ||
X56976.1 | - | 23 | 2819 |