Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287054.1 | NP_001273983.1 | 8 | 1771 | |
NM_001287055.1 | NP_001273984.1 | 4 | 1074 | ||
NM_003439.2 | NP_003430.1 | 6 | 1742 | ||
XM_011516559.2 | XP_011514861.1 | 6 | 2297 | ||
GenBank mRNA | AK054676.1 | - | 8 | 1744 | |
AK074116.1 | - | 6 | 1700 | ||
AK091471.1 | - | 4 | 1043 | ||
AK123012.1 | - | 1 | 1744 | ||
AL833578.1 | - | 6 | 1718 | ||
BC022378.1 | - | 6 | 1715 | 4685873 | |
BC112224.1 | - | 5 | 1545 | 8327692 | |
BC113697.1 | - | 5 | 1545 | 8322749 | |
BX640646.1 | - | 6 | 1715 | ||
U09848.1 | - | 2 | 785 |